missing translation for 'onlineSavingsMsg'
Learn More

Iron powder, 98% (metals basis), Thermo Scientific Chemicals

Product Code. 15421577
Click to view available options
Quantity:
250 g
500 g
1 kg
Unit Size:
1kg
250g
500g
This item is not returnable. View return policy

Product Code. 15421577

Brand: Thermo Scientific Alfa Aesar 039813.30

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7439-89-6
Molecular Formula Fe
Molecular Weight (g/mol) 55.85
InChI Key XEEYBQQBJWHFJM-UHFFFAOYSA-N
Synonym ferrum, iron, elemental, ferrous, remko, armco, ferrovac, hoeganaes eh, 3zhp, ancor b, atomiron 5m
PubChem CID 23925
ChEBI CHEBI:82664
IUPAC Name iron
SMILES [Fe]

Specifications

Color Gray
Appearance Gray powder
Density 7.87g/cm3 at 20°C
Quantity 250 g
Mesh Size -325 Mesh
Assay Percent Range 98% (Metals basis)
Form Powder
MDL Number MFCD00010999
Chemical Name or Material Iron, Reduced

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.