missing translation for 'onlineSavingsMsg'
Learn More

Iron Oxide (Solution in water), TRC

Product Code. 30398261
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30398261 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30398261 Supplier Toronto Research Chemicals Supplier No. TRCI2485405G
 Restricted Item
Estimated Shipment: 29-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Iron(III) Hydroxide Oxide
CAS 20344-49-4
Quantity 5 g
Molecular Formula Fe H O2
Synonym Iron Oxide, Iron hydroxide oxide (Fe(OH)O), Iron hydroxide oxide (FeO(OH)) (8CI), CFH 12, CFH 18, Colortherm 10, Colortherm Yellow 10, Colortherm Yellow C 10, Daipyroxide Yellow 8170, Depox, Duploxide 510P, E 331, E 331 (oxide), FEI 16PB, FeOOH, Ferric hydroxide oxide, Ferric hydroxide oxide (FeOOH), Ferric oxide hydroxide, Ferric oxyhydroxide (FeO2H), Ferric(III) acid, Ferronan 2505, Ferrosorp DGp, Ferrosorp DGp+K, Ferrosorp DGμ, Ferrous acid, GEH 101, Iron monohydroxide monooxide, Iron monoxide monohydroxide, Iron oxide hydroxide (FeO(OH)), Iron oxyhydroxide (FeO(OH)), Iron(III) oxide hydroxide, Iron(III) α-oxyhydroxide, LL 100P, Sicopur Yellow, Synthetic lepidocrocite, TSY 1, Tarox HY 250, Tarox LL 100P, Tarox LL-XLO, Tarox Synthetic Iron Oxide LL-XLO, Tarox XLO, Tarox Yellow LL 100P, Transyellow AC 500, Y 48, YLO 2288D, YM 1100, Yellow LL 100P, α-FeOOH, α-Iron hydroxide, α-Iron hydroxide oxide (FeOOH)
InChI Formula InChI=1S/Fe.H2O.O/h;1H2;/q+1;;/p-1
SMILES O[Fe]=O
IUPAC Name hydroxy(oxo)iron
Molecular Weight (g/mol) 88.852
Formula Weight 88.933
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.