missing translation for 'onlineSavingsMsg'
Learn More

Iron nitride

Product Code. 11309219
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11309219

Brand: Thermo Scientific Alfa Aesar 088198.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 37245-77-5 | Fe6H2N2-2 | 365.1 g/mol

Iron nitride is used in the synthesis of ammonia and carbon nanofibers. It is involved in the creation of ferrofluids, a liquid rocket fuel. It finds applications in making the strongest naturally magnetic material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 37245-77-5
Molecular Formula Fe6H2N2-2
Molecular Weight (g/mol) 365.1
MDL Number MFCD00134717
InChI Key ILMFDGFXPJCFQW-UHFFFAOYSA-N
Synonym iron nitride,-325 mesh powder, 1-aza-2,3,4-triferracyclobutan-1-yliron; diferrioamine
PubChem CID 56843031
IUPAC Name azanide;azanidylideneiron;iron
SMILES [NH2-].[N-]=[Fe].[Fe].[Fe].[Fe].[Fe].[Fe]

Specifications

Color Gray
Physical Form Powder
Quantity 100 g
Solubility Information Insoluble in water.
Odor Odorless
Appearance Gray powder
Chemical Name or Material Iron nitride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.