missing translation for 'onlineSavingsMsg'
Learn More

Iron Metal, Technical, Wire, about 0.3mm Diameter, Fisher Chemical

Product Code. 11448952 Shop All Fisher Chemical Products
Click to view available options
Quantity:
100 g
Packaging:
Plastic powder jar
Unit Size:
100g
This item is not returnable. View return policy

Product Code. 11448952

Brand: Fisher Chemical I/1001/48

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Color Gray
CAS 7439-89-6
Melting Point 1535°C
Boiling Point 3000°C
Quantity 100 g
Packaging Plastic powder jar
Vapor Pressure 1mmHg 1787
Form Solid
Molecular Formula Fe
MDL Number MFCD00010999
Synonym ferrum, iron, elemental, ferrous, remko, armco, ferrovac, hoeganaes eh, 3zhp, ancor b, atomiron 5m
InChI Key XEEYBQQBJWHFJM-UHFFFAOYSA-N
SMILES [Fe]
IUPAC Name iron
Molecular Weight (g/mol) 55.85
PubChem CID 23925
ChEBI CHEBI:82664
Formula Weight 55.84g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.