missing translation for 'onlineSavingsMsg'
Learn More

Iron Isotopic Standard: Fe @ 10 μg/mL in 2% HNO3 (Enriched in Fe-57), VHG™

Product Code. 17902886
Change view
Click to view available options
Quantity:
50 mL
Unit Size:
50mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17902886 50 mL 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17902886 Supplier VHG Labs Supplier No. VHGLIS57FE50
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

Iron Isotopic Standard: Fe @ 10 μg/mL in 2% HNO3 (Enriched in Fe-57)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Iron 57
CAS 14762-69-7
Methods ICP-MS
Analyte or Component Names Iron 57
Shipping Condition Room Temperature
Molecular Formula 57Fe
Synonym 57Fe
SMILES [57Fe]
IUPAC Name Iron 57
Solvent or Matrix Dilute HNO3
Molecular Weight (g/mol) 56.94
Quantity 50 mL
Formula Weight 56.9354 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.