missing translation for 'onlineSavingsMsg'
Learn More

Iron(III) trifluoromethanesulfonate, tech. 90%

Product Code. 15416508
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15416508 1 g 1g
15485426 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15416508 Supplier Thermo Scientific Alfa Aesar Supplier No. H60034.03
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 63295-48-7 | C3F9FeO9S3 | 503.035 g/mol

Iron(III) triflate is used as an efficient catalyst in the synthesis of 2,3-unsaturated-O-glycosides from 2,4,6-tri-O-acetyl-D-glucal. It is also involved in the synthesis of a variety of beta-enamino ketones and esters.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Iron(III) triflate is used as an efficient catalyst in the synthesis of 2,3-unsaturated-O-glycosides from 2,4,6-tri-O-acetyl-D-glucal. It is also involved in the synthesis of a variety of beta-enamino ketones and esters.

Solubility
Soluble in water, methanol and acetonitrile.

Notes
Moisture sensitive. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63295-48-7
Molecular Formula C3F9FeO9S3
Molecular Weight (g/mol) 503.035
MDL Number MFCD15144785
InChI Key OSHOQERNFGVVRH-UHFFFAOYSA-K
Synonym iron iii trifluoromethanesulfonate, fe otf 3, ferric triflate, iron iii triflate, fe cf3so3 3, ksc497q0p, iron 3+ tritriflate, ferric trifluoromethanesulfonate, iron 1,1,1-trifluoromethanesulfonic acid, iron 3+ ion tritrifluoromethanesulfonate
PubChem CID 11656316
IUPAC Name iron(3+);trifluoromethanesulfonate
SMILES C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].[Fe+3]

Specifications

Melting Point 183°C
Flash Point 110°C (230°F)
Linear Formula Fe(OSO2CF3)3
Quantity 1 g
Sensitivity Moisture sensitive
Solubility Information Soluble in water,methanol and acetonitrile.
Formula Weight 503.04
Percent Purity 90%
Grade Technical
Chemical Name or Material Iron(III) trifluoromethanesulfonate
compliance-icons
Product Identifier
  • Iron(III) trifluoromethanesulfonate
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to accordance with local/regional/national/international regulations..

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.