missing translation for 'onlineSavingsMsg'
Learn More

Iron(III) sulfate hydrate, Puriss. p.a., 21-23% Fe basis, Solstice

Product Code. 15623310
Change view
Click to view available options
Quantity:
500 g
1 kg
25 kg
6 x 1 kg
Packaging:
Paper / Poly Bag
Poly bottle
Unit Size:
1kg
25kg
500g
Pack of 6
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15623310 500 g Poly bottle 500g
15603310 1 kg Poly bottle 1kg
15633310 6 x 1 kg Poly bottle Pack of 6
15613310 25 kg Paper / Poly Bag 25kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15623310 Supplier Honeywell-Fluka Supplier No. 31235500G
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Puriss. p.a., 21-23% Fe basis

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15244-10-7
Molecular Formula Fe2H2O13S3
Molecular Weight (g/mol) 417.87
MDL Number MFCD00011007
InChI Key VXWSFRMTBJZULV-UHFFFAOYSA-H
PubChem CID 167265
IUPAC Name iron(3+);trisulfate;hydrate
SMILES O.[Fe+3].[Fe+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

Specifications

Quantity 500 g
Linear Formula Fe2(SO4)3 · xH2O
UN Number 3260
Formula Weight 399.88g/mol (anhydrous basis)
Grade puriss. p.a.
Concentration or Composition (by Analyte or Components) 21 to 23% Fe basis
Packaging Poly bottle
Chemical Name or Material Iron(Iii) Sulfate Hydrate
compliance-icons compliance-icons
Product Identifier
  • Iron(III) sulfate hydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H290-May be corrosive to metals.
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P234-Keep only in original container.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Iron(III) sulfate hydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.