missing translation for 'onlineSavingsMsg'
Learn More

Iron(III) sulfate hydrate, Puriss. p.a., 21-23% Fe basis, Honeywell Fluka™

Product Code. 15613310 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
500 g
1 kg
25 kg
6 x 1 kg
Packaging:
Paper / Poly Bag
Poly bottle
Unit Size:
1kg
25kg
500g
Pack of 6
This item is not returnable. View return policy

Product Code. 15613310

Brand: Honeywell-Fluka 3123525KG

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Puriss. p.a., 21-23% Fe basis

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15244-10-7, 7664-93-9
Molecular Formula Fe2H2O13S3
Molecular Weight (g/mol) 417.87
MDL Number MFCD00011007
InChI Key VXWSFRMTBJZULV-UHFFFAOYSA-H
Synonym Ferric sulfate hydrate
PubChem CID 167265
IUPAC Name iron(3+);trisulfate;hydrate
SMILES O.[Fe+3].[Fe+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

Specifications

CAS Max % 98.0000%, 2.0000%
Quantity 25 kg
Linear Formula Fe2(SO4)3 · xH2O
UN Number UN3260
Formula Weight 399.88
Grade puriss. p.a.
Packaging Paper / Poly Bag
Chemical Name or Material Iron(III) sulfate hydrate, Puriss. p.a., 21-23% Fe basis
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.