missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iron(Iii) Sulfate Hydrate, Honeywell Fluka™
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
| Quantity | 100 g |
| Linear Formula | Fe2(SO4)3 · xH2O |
| UN Number | 3260 |
| Formula Weight | 399.88g/mol (anhydrous basis) |
| Percent Purity | 97% |
| Chemical Name or Material | Iron(Iii) Sulfate Hydrate |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction