missing translation for 'onlineSavingsMsg'
Learn More

Iron(III) phosphate hydrate, extra pure

Product Code. 10433545 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Glass bottle
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10433545 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10433545 Supplier Thermo Scientific Acros Supplier No. 389955000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 51833-68-2 | FeH2O5P | 168.83 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 51833-68-2
Color White
Physical Form Powder, Crystals, Granules and/or Chunks
Quantity 500 g
Molecular Formula FeH2O5P
Linear Formula FePO4·xH2O
MDL Number MFCD00150236
Synonym ferric phosphate hydrate, iron iii phosphate hydrate, ferric orthophosphate hydrate, ccris 8095, acmc-20akpj, fe.po4.h2o, ksc497s7j, iron 3+ hydrate phosphate, iron 3+ ion hydrate phosphate, phosphoric acid, iron 3 salt 1:1 , hydrate
Solubility Information Solubility in water: insoluble
InChI Key RQKNQWUJROKKEQ-UHFFFAOYSA-K
SMILES O.[Fe+3].[O-]P([O-])([O-])=O
IUPAC Name iron(3+) hydrate phosphate
Molecular Weight (g/mol) 168.83
PubChem CID 21410239
Formula Weight 150.82
Percent Purity 26.0 to 32.0% (as Fe)
Grade Extra Pure
Loss on Ignition 32.5 % max.
Packaging Glass bottle
Chemical Name or Material Iron(III) phosphate hydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.