Learn More
Iron(III) perchlorate hydrate, Reagent Grade, Thermo Scientific Chemicals
Description
Iron(III) perchlorate hydrate is used as a starting material for the synthesis of a tris-salicylate Fe(III) complex having bidentate chelating units that allow two coordination modes. It is also used as a catalyst for synthesis of fullerene-fused lactones, α-Carbonyl furans via one-pot cyclization, fullerodioxolanes via heterocyclization, dialylated indoles via double alkylation, aryl esters via oxidative esterification and as a chemical actinometer based on photolysis of ferrioxalate in presence of polyoxometalate in aqueous solution, as an oxidant for conducting polymer nanoparticles synthesized in ionic liquid by chemical polymerization.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | White |
Physical Form | Crystalline |
Quantity | 100 g |
Solubility Information | Soluble in water. |
Formula Weight | 354.19 (Anhydrous) |
Grade | Reagent |
Odor | Odorless |
Sensitivity | Hygroscopic |
Chemical Name or Material | Iron(III) perchlorate hydrate |
Safety and Handling
- Iron(II) perchlorate hydrate
- Danger
- Oxidising solid Category 2
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- H272-May intensify fire; oxidiser.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P220-Keep/Store away from clothing/combustible materials.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P501-Dispose of contents/container to .
RUO – Research Use Only