missing translation for 'onlineSavingsMsg'
Learn More

Iron(Iii) Oxide, puriss., Honeywell Fluka™

Product Code. 15651130 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
1 kg
Unit Size:
1kg
This item is not returnable. View return policy

Product Code. 15651130

Brand: Honeywell-Fluka 123441KG

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 1309-37-1
Quantity 1 kg
Molecular Formula Fe2O3
MDL Number MFCD00011008
UN Number NONH for all modes of transport
InChI Key JEIPFZHSYJVQDO-UHFFFAOYSA-N
SMILES [Fe][Fe](=O)(=O)=O
IUPAC Name trioxodiiron
Molecular Weight (g/mol) 159.69
Formula Weight 159.69g/mol
Percent Purity ≥97% (complexometric)
Grade puriss.
Chemical Name or Material Iron(Iii) Oxide
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.