missing translation for 'onlineSavingsMsg'
Learn More

Iron(III) oxide, cosmetic, NanoArc™, Thermo Scientific Chemicals

Product Code. 11450870
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11450870 500 g 500g
15422067 25 g 25g
11440870 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11450870 Supplier Thermo Scientific Alfa Aesar Supplier No. 044895.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Nanoarc Cosmetic Iron Oxide Is An Auburn Material offering Unique Cosmetic Qualitites. The Small Particle Size Results in a Strong Color Development Combined With High Translucency, For a Unique Glowing Look

NanoArc cosmetic iron oxide is an auburn material offering unique cosmetic qualities. The small particle size results in a strong color development combined with high translucency for a unique glowing look.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

S.A. 30-60m2/g
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1309-37-1
Molecular Formula Fe2O3
Molecular Weight (g/mol) 159.69
MDL Number MFCD00011008
InChI Key JEIPFZHSYJVQDO-UHFFFAOYSA-N
IUPAC Name trioxodiiron
SMILES [Fe][Fe](=O)(=O)=O

Specifications

Melting Point 1565°C
Color Red to Brown
Physical Form Powder
Quantity 500 g
Solubility Information Insoluble in water
Formula Weight 159.69
Odor Odorless
Appearance Red-brown powder
Density 5.24
Chemical Name or Material Iron(III) oxide, cosmetic, Sold in collaboration with Nanophase Technologies Corporation

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.