Learn More
Iron(III) oxide, cosmetic, NanoArc™, Thermo Scientific Chemicals
Description
NanoArc cosmetic iron oxide is an auburn material offering unique cosmetic qualities. The small particle size results in a strong color development combined with high translucency for a unique glowing look.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 1565°C |
| Color | Red to Brown |
| Physical Form | Powder |
| Quantity | 100 g |
| Solubility Information | Insoluble in water |
| Formula Weight | 159.69 |
| Odor | Odorless |
| Appearance | Red-brown powder |
| Density | 5.24 g/cm3 at 20°C |
| Chemical Name or Material | Iron(III) oxide, cosmetic, Sold in collaboration with Nanophase Technologies Corporation |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.