missing translation for 'onlineSavingsMsg'
Learn More

Iron(III) oxide, alpha-phase, nanopowder, 98% (metals basis), Thermo Scientific Chemicals

Product Code. 15427375
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15427375 25 g 25g
15437375 100 g 100g
15447375 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15427375 Supplier Thermo Scientific Alfa Aesar Supplier No. 047044.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Ferric oxide is used for polishing precious metals and diamonds, as well as in cosmetics. Synthetic iron oxide pigments are one of the most popular color pigments worldwide for coloring concrete. Other uses includes: coating, printing ink, paint, also as coloring agent for building material, rubber, paper-making. In producing ferrites . Also used in recording tapes and pyrotechnics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1309-37-1
Molecular Formula Fe2O3
Molecular Weight (g/mol) 159.69
MDL Number MFCD00011008
InChI Key JEIPFZHSYJVQDO-UHFFFAOYSA-N
IUPAC Name trioxodiiron
SMILES [Fe][Fe](=O)(=O)=O

Specifications

Melting Point 1565°C
Color Red to Brown
Physical Form Powder
Quantity 25 g
Merck Index 14,4028
Solubility Information It is soluble In Warm Hydrochloric Acid,Slightly Soluble in Sulfuric Acid.
Formula Weight 159.69
Percent Purity 98%
Odor Odorless
Density 5.24 g/mL
Assay (metals basis)
Chemical Name or Material Iron (III) oxide, alpha-phase
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.