missing translation for 'onlineSavingsMsg'
Learn More

Iron(Iii) Oxide, 99.98% trace metals basis, Honeywell™

Product Code. 15611750 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
10 g
Packaging:
Poly Bottle
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15611750 10 g Poly Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15611750 Supplier Honeywell Research Chemicals Supplier No. 20351310G

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Ver productos alternativos

missing translation for 'validMainframeMsg'

Este artículo no se puede devolver. Vea la política de devoluciones

99.98% trace metals basis

  • We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
    Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
    Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
TRUSTED_SUSTAINABILITY

Especificaciones

Melting Point 1565°C
CAS 1309-37-1
Color Red
Physical Form Solid
Quantity 10 g
Molecular Formula Fe2O3
MDL Number MFCD00011008
UN Number NONH for all modes of transport
InChI Key JEIPFZHSYJVQDO-UHFFFAOYSA-N
SMILES [Fe][Fe](=O)(=O)=O
IUPAC Name trioxodiiron
Molecular Weight (g/mol) 159.69
Formula Weight 159.69g/mol
Percent Purity 99.98%
Packaging Poly Bottle
Density 5g/cu.cm
Chemical Name or Material Iron(Iii) Oxide
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.