missing translation for 'onlineSavingsMsg'
Learn More

Iron(III) oxide, 99.9% (metals basis), Thermo Scientific Chemicals

Product Code. 15492917
Change view
Click to view available options
Quantity:
250 g
Unit Size:
250g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15492917 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15492917 Supplier Thermo Scientific Alfa Aesar Supplier No. 047247.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Iron(III) oxide acts as a pigment in paints, coatings and colored concretes, used in thermalite. Iron oxide is a low cost, nontoxic, environmental friendly catalyst for various reactions such as oxidation of carbon monoxide, reduction of nitrogen oxides and sulfur dioxide by CO, photocatalytic splitting of water, Fischer-Tropsch synthesis of hydrocarbons, catalytic decomposition of industrial dyes, water-gas shift reactions, nucleation of diamonds, conversion of phenolic and other environmentally harmful aromatic compounds, dehydrogenation of ethylbenzene to styrene, and catalytic oxidations of various organic compounds. It is used in magnetic storage devices and in the production of iron, steel and alloys. It functions as a catalyst in the chemical oxidation of hydrogen peroxide used for the treatment of contaminated water with hydrogen peroxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1309-37-1
Molecular Formula Fe2O3
Molecular Weight (g/mol) 159.69
MDL Number MFCD00011008
InChI Key JEIPFZHSYJVQDO-UHFFFAOYSA-N
IUPAC Name trioxodiiron
SMILES [Fe][Fe](=O)(=O)=O

Specifications

Melting Point 1565°C
Physical Form Powder
Quantity 250 g
Merck Index 14,4028
Solubility Information Insoluble in water. Soluble in dilute acids.
Formula Weight 159.69
Percent Purity 99.9%
Odor Odorless
Assay (metals basis)
Chemical Name or Material Iron(III) oxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.