missing translation for 'onlineSavingsMsg'
Learn More

Iron(III) oxide, 95%, pure

Product Code. p-7077751 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 kg
5 kg
25 kg
Unit Size:
1kg
25kg
5kg
This item is not returnable. View return policy

Product Code. 15582343

missing translation for 'mfr': Thermo Scientific Acros 458700010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1309-37-1 | Fe2O3 | 159.69 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1309-37-1
Molecular Formula Fe2O3
Molecular Weight (g/mol) 159.69
MDL Number MFCD00011008
InChI Key JEIPFZHSYJVQDO-UHFFFAOYSA-N
Synonym Ferric oxide
IUPAC Name trioxodiiron
SMILES [Fe][Fe](=O)(=O)=O

Specifications

Melting Point 1538.0°C
Quantity 1 kg
Assay Percent Range 94% min. (Iodometry)
Linear Formula Fe2O3
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in acids, soluble in alcohol and ether
Formula Weight 159.69
Percent Purity 95%
Grade Pure
Loss on Drying 0.3% max.
Specific Gravity 5.24
Density 5.2400g/mL
Chemical Name or Material Iron(III) oxide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.