missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iron(III) oxalate hexahydrate, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
Iron(III) oxalate hexahydrate is used as a catalyst for the reductive and oxidative degradation of iopamidol under UV and visible light irradiation.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Quantity | 25 g |
UN Number | UN3288 |
Solubility Information | Soluble in water. |
Formula Weight | 483.84 (375.55 Anhydrous) |
Physical Form | Powder |
Chemical Name or Material | Iron(III) oxalate hexahydrate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'