missing translation for 'onlineSavingsMsg'
Learn More

Iron(Iii) Nitrate Nonahydrate, Solstice

Product Code. 15652040 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
100 g
250 g
500 g
1 kg
2.5 kg
Unit Size:
100g
1kg
2.5kg
250g
500g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15652040 100 g 100g
15682040 250 g 250g
15692040 500 g 500g
15662040 1 kg 1kg
15672040 2.5 kg 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 15652040 Supplier Honeywell-Fluka Supplier No. 216828100G
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

ACS Reagent,≥98%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7782-61-8
Molecular Formula FeH18N3O18
Molecular Weight (g/mol) 403.99
MDL Number MFCD00149708
InChI Key SZQUEWJRBJDHSM-UHFFFAOYSA-N
Synonym ferric nitrate nonahydrate, iron iii nitrate nonahydrate, unii-vmx4uop3vn, ferric nitrate, nonahydrate, iron iii nitrate, vmx4uop3vn, iron, reference standard solution, ferricnitratenonahydrate, nitric acid, iron 3+ salt, nonahydrate, iron iii nitrate hydrate
PubChem CID 16211566
IUPAC Name iron(3+);trinitrate;nonahydrate

Specifications

Quantity 100 g
Linear Formula Fe(NO3)3 · 9H2O
UN Number 3260
SMILES O.O.O.O.O.O.O.O.O.[Fe+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
Formula Weight 404g/mol
Percent Purity ≥98%
Grade ACS Reagent
Chemical Name or Material Iron(Iii) Nitrate Nonahydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.