missing translation for 'onlineSavingsMsg'
Learn More

Iron(III) Nitrate Nonahydrate, Certified AR for Analysis, Fisher Chemical™

Product Code. p-7073098 Shop All Fisher Chemical Products
Click to view available options
Quantity:
250 g
500 g
1 kg
Unit Size:
1kg
250g
500g
This item is not returnable. View return policy

Product Code. 10171520

Brand: Fisher Chemical I/1075/50

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Iron(III) Nitrate Nonahydrate, Certified AR for Analysis

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7782-61-8
Molecular Formula FeH18N3O18
Molecular Weight (g/mol) 403.99
MDL Number MFCD00149708
InChI Key SZQUEWJRBJDHSM-UHFFFAOYSA-N
Synonym ferric nitrate nonahydrate, iron iii nitrate nonahydrate, unii-vmx4uop3vn, ferric nitrate, nonahydrate, iron iii nitrate, vmx4uop3vn, iron, reference standard solution, ferricnitratenonahydrate, nitric acid, iron 3+ salt, nonahydrate, iron iii nitrate hydrate
PubChem CID 16211566
SMILES O.O.O.O.O.O.O.O.O.[Fe+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Melting Point 47°C
Color Purple
Physical Form Solid
Quantity 250 g
IUPAC Name iron(3+) nonahydrate trinitrate
Formula Weight 404g/mol
Packaging Plastic powder jar
compliance-icons compliance-icons
Product Identifier
  • Iron(III) nitrate nonahydrate
Signal Word
  • Warning
Hazard Category
  • Oxidising solid Category 3
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : Iron(III) nitrate nonahydrate, Ferric nitrate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.