missing translation for 'onlineSavingsMsg'
Learn More

Iron(III) nitrate nonahydrate, 99.9995%, (trace metal basis)

Product Code. 10492570 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10492570 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10492570 Supplier Thermo Scientific Acros Supplier No. 436810250
 Restricted Item
Estimated Shipment: 29-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 7782-61-8 | FeH18N3O18 | 403.99 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis
TRUSTED_SUSTAINABILITY

Specifications

Melting Point 47.0°C
CAS 7782-61-8
Color Purple to White or Violet
Physical Form Crystals
Quantity 25 g
Assay Percent Range 13.5 to 14.2% (Fe) (Iodometry)
Molecular Formula FeH18N3O18
Linear Formula Fe(NO3)3·9H2O
MDL Number MFCD00149708
Merck Index 15,4054
Synonym ferric nitrate nonahydrate, iron iii nitrate nonahydrate, unii-vmx4uop3vn, ferric nitrate, nonahydrate, iron iii nitrate, vmx4uop3vn, iron, reference standard solution, ferricnitratenonahydrate, nitric acid, iron 3+ salt, nonahydrate, iron iii nitrate hydrate
Solubility Information Solubility in water: soluble. Other solubilities: very soluble in ethanol, acetone
InChI Key SZQUEWJRBJDHSM-UHFFFAOYSA-N
SMILES O.O.O.O.O.O.O.O.O.[Fe+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
IUPAC Name iron(3+);trinitrate;nonahydrate
Molecular Weight (g/mol) 403.99
PubChem CID 16211566
Formula Weight 404
Percent Purity 99.9995%
Grade Trace Metal Basis
Packaging Glass bottle
Specific Gravity 1.68
Density 1.6800g/mL
Chemical Name or Material Iron(III) nitrate nonahydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.