missing translation for 'onlineSavingsMsg'
Learn More

Iron(III) nitrate nonahydrate, 98+%, ACS reagent

Product Code. 10301741 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 g
3 kg
Packaging:
Plastic Bottle
Unit Size:
3kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10301741 3 kg Plastic Bottle 3kg
10596982 500 g Plastic Bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10301741 Supplier Thermo Scientific Acros Supplier No. 423710030
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7782-61-8 | FeH18N3O18 | 403.99 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7782-61-8
Molecular Formula FeH18N3O18
Molecular Weight (g/mol) 403.99
MDL Number MFCD00149708
InChI Key SZQUEWJRBJDHSM-UHFFFAOYSA-N
Synonym ferric nitrate nonahydrate, iron iii nitrate nonahydrate, unii-vmx4uop3vn, ferric nitrate, nonahydrate, iron iii nitrate, vmx4uop3vn, iron, reference standard solution, ferricnitratenonahydrate, nitric acid, iron 3+ salt, nonahydrate, iron iii nitrate hydrate
PubChem CID 16211566
IUPAC Name iron(3+) nonahydrate trinitrate
SMILES O.O.O.O.O.O.O.O.O.[Fe+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Melting Point 47.0°C
Color Gray to Purple
Physical Form Adhering Crystalline Powder
Quantity 3 kg
Assay Percent Range 98.0 to 101% (Complexometry)
Linear Formula Fe(NO3)3·9H2O
UN Number 1466
Merck Index 15, 4054
Solubility Information Solubility in water: soluble. Other solubilities: very soluble in ethanol, acetone
Formula Weight 404
Percent Purity ≥98%
Grade ACS Reagent
Packaging Plastic Bottle
Specific Gravity 1.68
Vapor Pressure Negligible
Decomposition Information Decomposition Information: < 100°C
Density 1.6800g/mL
Chemical Name or Material Iron(III) nitrate nonahydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.