missing translation for 'onlineSavingsMsg'
Learn More

Iron(III) meso-tetraphenylporphine chloride

Product Code. 11393487
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11393487 1 g 1g
11303497 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11393487 Supplier Thermo Scientific Alfa Aesar Supplier No. 018829.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 16456-81-8 | C44H30Cl3FeN4 | 776.947 g/mol

Iron(III) meso-tetraphenylporphine chloride is used catalyzing reagent for silylation reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16456-81-8
Molecular Formula C44H30Cl3FeN4
Molecular Weight (g/mol) 776.947
MDL Number MFCD00012152
InChI Key QDTNPIPZGCOFPA-UHFFFAOYSA-K
Synonym 5,10,15,20-tetraphenyl-21h,23h-porphine iron iii chloride, acmc-20am7m, chloro 5,10,15,20-tetraphenyl porphyrinato iron iii, iron iii meso-tetraphenylporphine chloride
PubChem CID 67043998
IUPAC Name iron(3+);5,10,15,20-tetraphenyl-21,22-dihydroporphyrin;trichloride
SMILES C1=CC=C(C=C1)C2=C3C=CC(=C(C4=CC=C(N4)C(=C5C=CC(=N5)C(=C6C=CC2=N6)C7=CC=CC=C7)C8=CC=CC=C8)C9=CC=CC=C9)N3.[Cl-].[Cl-].[Cl-].[Fe+3]

Specifications

Physical Form Crystalline
Quantity 1 g
Solubility Information Insoluble in water.
Formula Weight 704.04
Chemical Name or Material Iron(III) meso-tetraphenylporphine chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.