missing translation for 'onlineSavingsMsg'
Learn More

Iron(III) meso-Tetraphenylporphine Chloride, TRC

Product Code. 30403071
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30403071 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30403071 Supplier Toronto Research Chemicals Supplier No. TRCI775205250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Iron(III) meso-Tetraphenylporphine Chloride
CAS 16456-81-8
Molecular Formula C44 H28 Cl Fe N4
Quantity 250 mg
Synonym [alpha,beta,gamma,delta-Tetraphenylporphinato(2-)]iron Chloride, (SP-5-12)-Chloro[5,10,15,20-tetraphenyl-21H,23H-porphinato(2-)-kN21,kN22,kN23,kN24]iron, (5,10,15,20-Tetraphenylporphinato)iron(III) Chloride, 5,10,15,20-Tetraphenyl-21H,23H-porphine iron(III) Chloride, 5,10,15,20-Tetraphenylporphyrinatoiron(III) Chloride, 5,10,15,20-Tetraphenylporphyriniron(III) Chloride, Chloro(5,10,15,20-tetraphenylporphinato)iron, Chloro(5,10,15,20-tetraphenylporphyrin)iron, Chloro(5,10,15,20-tetraphenylporphyrinato)iron(III), Chloro(meso-tetraphenylporphinato)iron, Chloro(meso-tetraphenylporphyrinato)iron, Chloro(tetraphenylporphinato)iron, Chloro(tetraphenylporphyrinato)iron, Chlorotetraphenylporphinatoiron(III), Iron Chlorotetraphenylporphyrin, Iron meso-Tetraphenylporphyrin Chloride, Iron Tetraphenylporphine Chloride, Iron Tetraphenylporphyrin Chloride, Iron(5,10,15,20-tetraphenylporphyrin) Chloride, Iron(III) Tetraphenylporphine Chloride, Iron(III) Tetraphenylporphyrin Chloride, Mesotetraphenylporphyrin Iron(III) Chloride, Tetraphenylporphinatoiron(III) Chloride, Tetraphenylporphineiron(III) Chloride, Tetraphenylporphyrin Iron(III) Chloride, [5,10,15,20-Tetraphenylporphinato(2-)]iron(1+) Chloride, [alpha,beta,gamma,delta-Tetraphenylporphyrinato(2-)]iron(1+) Chloride
InChI Formula InChI=1S/C44H28N4.ClH.Fe/c1-5-13-29(14-6-1)41-33-21-23-35(45-33)42(30-15-7-2-8-16-30)37-25-27-39(47-37)44(32-19-11-4-12-20-32)40-28-26-38(48-40)43(31-17-9-3-10-18-31)36-24-22-34(41)46-36;;/h1-28H;1H;/q-2;;+3/p-1/b41-33-,41-34-,42-35-,42-37-,43-36-,43-38-,44-39-,44-40-;;
SMILES [Cl-][Fe+3]123[N]4=C5C=CC4=C(C6=CC=C(C(=C7C=CC(=[N]17)C(=C8C=CC(=C5c9ccccc9)[N-]28)c%10ccccc%10)c%11ccccc%11)[N-]36)c%12ccccc%12
Molecular Weight (g/mol) 704.02
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.