missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iron(III) meso-Tetraphenylporphine Chloride, TRC
Specifications
Specifications
| Chemical Name or Material | Iron(III) meso-Tetraphenylporphine Chloride |
| CAS | 16456-81-8 |
| Molecular Formula | C44 H28 Cl Fe N4 |
| Quantity | 50 mg |
| Synonym | [alpha,beta,gamma,delta-Tetraphenylporphinato(2-)]iron Chloride, (SP-5-12)-Chloro[5,10,15,20-tetraphenyl-21H,23H-porphinato(2-)-kN21,kN22,kN23,kN24]iron, (5,10,15,20-Tetraphenylporphinato)iron(III) Chloride, 5,10,15,20-Tetraphenyl-21H,23H-porphine iron(III) Chloride, 5,10,15,20-Tetraphenylporphyrinatoiron(III) Chloride, 5,10,15,20-Tetraphenylporphyriniron(III) Chloride, Chloro(5,10,15,20-tetraphenylporphinato)iron, Chloro(5,10,15,20-tetraphenylporphyrin)iron, Chloro(5,10,15,20-tetraphenylporphyrinato)iron(III), Chloro(meso-tetraphenylporphinato)iron, Chloro(meso-tetraphenylporphyrinato)iron, Chloro(tetraphenylporphinato)iron, Chloro(tetraphenylporphyrinato)iron, Chlorotetraphenylporphinatoiron(III), Iron Chlorotetraphenylporphyrin, Iron meso-Tetraphenylporphyrin Chloride, Iron Tetraphenylporphine Chloride, Iron Tetraphenylporphyrin Chloride, Iron(5,10,15,20-tetraphenylporphyrin) Chloride, Iron(III) Tetraphenylporphine Chloride, Iron(III) Tetraphenylporphyrin Chloride, Mesotetraphenylporphyrin Iron(III) Chloride, Tetraphenylporphinatoiron(III) Chloride, Tetraphenylporphineiron(III) Chloride, Tetraphenylporphyrin Iron(III) Chloride, [5,10,15,20-Tetraphenylporphinato(2-)]iron(1+) Chloride, [alpha,beta,gamma,delta-Tetraphenylporphyrinato(2-)]iron(1+) Chloride |
| InChI Formula | InChI=1S/C44H28N4.ClH.Fe/c1-5-13-29(14-6-1)41-33-21-23-35(45-33)42(30-15-7-2-8-16-30)37-25-27-39(47-37)44(32-19-11-4-12-20-32)40-28-26-38(48-40)43(31-17-9-3-10-18-31)36-24-22-34(41)46-36;;/h1-28H;1H;/q-2;;+3/p-1/b41-33-,41-34-,42-35-,42-37-,43-36-,43-38-,44-39-,44-40-;; |
| SMILES | [Cl-][Fe+3]123[N]4=C5C=CC4=C(C6=CC=C(C(=C7C=CC(=[N]17)C(=C8C=CC(=C5c9ccccc9)[N-]28)c%10ccccc%10)c%11ccccc%11)[N-]36)c%12ccccc%12 |
| Molecular Weight (g/mol) | 704.02 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?