missing translation for 'onlineSavingsMsg'
Learn More

Iron(III) hydroxide, gamma-phase

Product Code. p-4904543
Click to view available options
Quantity:
2 g
10 g
50 g
Unit Size:
10g
2g
50g
This item is not returnable. View return policy

Product Code. 11343367

Brand: Thermo Scientific Alfa Aesar 017531.04

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 20344-49-4 | FeHO2 | 88.851 g/mol

Iron(III) hydroxide, γ-phase is used in purifying water and as an absorbent in chemical processes. It is involved in the preparation of iron oxide-hydroxides nanoparticles, which is used as a very good adsorbent for lead removal from aquatic media. It is also used as a pigment, as a catalyst and in aquarium water treatment as a phosphate binder. Further, it is employed in the paints-lacquers-varnishes industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20344-49-4
Molecular Formula FeHO2
Molecular Weight (g/mol) 88.851
MDL Number MFCD00064782
InChI Key AEIXRCIKZIZYPM-UHFFFAOYSA-M
Synonym hydroxy oxo iron, goethite, lepidocrocite, iron hydroxide oxide, goethite fe oh o, iron hydroxide oxide fe oh o, iron iii oxide-hydroxide, ferric acid, iron pigment yellow, ferric hydroxide oxide
PubChem CID 91502
IUPAC Name hydroxy(oxo)iron
SMILES O[Fe]=O

Specifications

Melting Point 135°C (decomposition)
Physical Form Powder
Quantity 2 g
Merck Index 14,4025
Solubility Information Insoluble in water.
Formula Weight 88.85
Odor Odorless
Chemical Name or Material Iron(III) hydroxide, gamma-phase

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.