missing translation for 'onlineSavingsMsg'
Learn More

Iron(III) hydroxide, 20% in H{2}O, nanoparticle dispersion, low pH, <5.0nm APS Number Weighted

Product Code. p-7052347
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15435085 100 g 100g
15445085 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15435085 Supplier Thermo Scientific Alfa Aesar Supplier No. 047241.22
Add to basket
This item is not returnable. View return policy

CAS: 20344-49-4 | FeHO2 | 88.851 g/mol

Iron(III) hydroxide is used in cosmetics, tattoo inks and aquarium water treatment. It is used as a pigment and as an adsorbent for removing lead from aquatic media. It acts as a catalyst for proteomics research. It is involved in the removal of toxic selenate and selenite ions from water systems.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20344-49-4
Molecular Formula FeHO2
Molecular Weight (g/mol) 88.851
MDL Number MFCD00064782
InChI Key AEIXRCIKZIZYPM-UHFFFAOYSA-M
Synonym hydroxy oxo iron, goethite, lepidocrocite, iron hydroxide oxide, goethite fe oh o, iron hydroxide oxide fe oh o, iron iii oxide-hydroxide, ferric acid, iron pigment yellow, ferric hydroxide oxide
PubChem CID 91502
IUPAC Name hydroxy(oxo)iron
SMILES O[Fe]=O

Specifications

Color Red to Brown
pH 3.0
Physical Form Liquid
Quantity 100 g
Linear Formula FeO(OH)
Merck Index 14,4025
Solubility Information Insoluble in water.
Formula Weight 88.85
Odor Odorless
Density 1.20 g/mL
Chemical Name or Material Iron(III) hydroxide, 20% in H2O, nanoparticle dispersion
Show More Show Less
compliance-icons
Product Identifier
  • Iron(III) hydroxide, 20% in H2O, nanoparticle dispersion, low pH
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P362-Take off contaminated clothing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to accordance with local/regional/national/international regulations..

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.