missing translation for 'onlineSavingsMsg'
Learn More

Iron(III) fluoride, anhydrous, 97% min

Product Code. p-4407480
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 11307157

Brand: Thermo Scientific Alfa Aesar 011487.09

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7783-50-8 | F3Fe | 112.84 g/mol

Iron(III) fluroide is used in ceramics manufacturing. It is used as a catalyst for certain cross coupling reactions. Further, it is involved in the chemoselective addition of cyanide to aldehyde to prepare cyanohydrins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7783-50-8
Molecular Formula F3Fe
Molecular Weight (g/mol) 112.84
MDL Number MFCD00011012
InChI Key SHXXPRJOPFJRHA-UHFFFAOYSA-K
Synonym ferric fluoride, iron iii fluoride, iron fluoride, iron trifluoride, iron fluoride fef3, ferric trifluoride, fef3, hsdb 450, iron iii fluoride, anhydrous, ferric fluoride, anhydrous
PubChem CID 24552
IUPAC Name trifluoroiron
SMILES F[Fe](F)F

Specifications

Melting Point >1000° (sublimation)
Color Green
Physical Form Powder
Quantity 10 g
Assay Percent Range 97% min. (Metal basis)
Solubility Information Soluble in dilute hydrofluroic acid. Slightly soluble in water. Insoluble in alcohol,ether and benzene.
Formula Weight 112.84
Sensitivity Hygroscopic
Appearance Pale green powder
Density 3.87 g/cm3
Chemical Name or Material Iron(III) fluoride
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Iron(II) fluoride, anhydrous
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 1 C
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.