missing translation for 'onlineSavingsMsg'
Learn More

Iron(III) Ferrocyanide, TRC

Product Code. 30399807
Change view
Click to view available options
Quantity:
250 mg
500 mg
2.5 g
Unit Size:
2.5g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30399807 500 mg -
30389914 2.5 g 2.5g
30407352 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30399807 Supplier Toronto Research Chemicals Supplier No. TRCI775228500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14038-43-8
Molecular Formula C18Fe7N18
Molecular Weight (g/mol) 859.23
Synonym (OC-6-11)-hexakis(cyano-kC)-Ferrate(4-) iron(3+), hexacyano-Ferrate(4-) iron(3+), (OC-6-11)-hexakis(cyano-C)-Ferrate(4-) iron(3+), Iron ferrocyanide (Fe4[Fe(CN)6]3), Antidotum-Thallii-Heyl, CI 77510 Blue, hexakis(cyano-C)di-Ferrate(1-), Ferric ferrocyanide, Ferric ferrocyanide (Fe4[Fe(CN)6]3), Ferric hexacyanoferrate, Ferric hexacyanoferrate (II), Ferrihexacyanoferrate, Ferrocin, Ferrotsin, Iron ferrocyanide, Iron(3+) ferrocyanide, Iron(3+) hexacyanoferrate(4-) (4:3), Milori blue, NSC 8665, Prussian blue, Prussian blue [Fe4[Fe(CN)6]3], Prussian blue insoluble, Radiogardase Cs, Tetrairon tris(hexacyanoferrate(4-)), Tetrairon tris(hexacyanoferrate), Tetrairon(3+) tris[hexacyanoferrate(4-)]
IUPAC Name hexakis(azanylidynemethyl)iron(2+);iron(3+)
SMILES [Fe+3].[Fe+3].[Fe+3].[Fe+3].N#[C-][Fe+2]([C-]#N)([C-]#N)([C-]#N)([C-]#N)[C-]#N.N#[C-][Fe+2]([C-]#N)([C-]#N)([C-]#N)([C-]#N)[C-]#N.N#[C-][Fe+2]([C-]#N)([C-]#N)([C-]#N)([C-]#N)[C-]#N

Specifications

Quantity 500 mg
InChI Formula InChI=1S/18CN.7Fe/c18*1-2;;;;;;;/q18*-1;3*+2;4*+3
Formula Weight 859.5999
Chemical Name or Material Iron(III) Ferrocyanide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.