missing translation for 'onlineSavingsMsg'
Learn More

Iron(III) Ferrocyanide, Honeywell™ Fluka™

Product Code. 15672320 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15672320 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15672320 Supplier Honeywell-Fluka Supplier No. 23412525G
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Powder,≥30.0 % Iron (Fe) by ICP Assay

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Specifications

CAS 14038-43-8
Color Blue
Physical Form Powder
Quantity 25 g
Molecular Formula C18Fe7N18
Linear Formula Fe4[Fe(CN)6]3
MDL Number MFCD00135663
UN Number NONH for all modes of transport
Synonym prussian blue, ferric ferrocyanide, iron iii ferrocyanide, iron iii hexacyanoferrate ii, ferric hexacyanoferrate, parisian blue, berlin blue, ferrocin, preussischblau, turnbulls blau
InChI Key DCYOBGZUOMKFPA-UHFFFAOYSA-N
SMILES [C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[Fe+2].[Fe+2].[Fe+2].[Fe+3].[Fe+3].[Fe+3].[Fe+3]
IUPAC Name iron(2+);iron(3+);octadecacyanide
Molecular Weight (g/mol) 859.239
PubChem CID 2724251
Formula Weight 859.23g/mol
Packaging Glass bottle
Chemical Name or Material Iron(Iii) Ferrocyanide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.