missing translation for 'onlineSavingsMsg'
Learn More

Iron(III) ethoxide, 99.6% (metals basis)

Product Code. 11364837
Change view
Click to view available options
Quantity:
0.5 g
2 g
10 g
50 g
Unit Size:
0.5g
10g
2g
50g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11364837 2 g 2g
11394837 0.5 g 0.5g
11374837 10 g 10g
11384837 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11364837 Supplier Thermo Scientific Alfa Aesar Supplier No. 041863.04
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Precursor for Sol-Gel processes | CAS: 5058-42-4 | C6H15FeO3 | 191.028 g/mol

Precursor for Sol-Gel processes.Iron(III) ethoxide is used as a precursor for inorganic compound iron oxide. It is used in the preparation of iron-platinum nanoparticles by pyrolysis with platinum(II) acetylacetonate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5058-42-4
Molecular Formula C6H15FeO3
Molecular Weight (g/mol) 191.028
MDL Number MFCD00078028
InChI Key JTPUGUWXHGEEHW-UHFFFAOYSA-N
Synonym iron iii ethoxide, iron 3+ ethanolate, triethoxyiron iii, ethanolate; iron 3+, acmc-1alnr, iron 3+ tris ethoxide, iron 3+ ion tris ethoxide, iron iii ethoxide trace metals basis 500mg
PubChem CID 6452222
IUPAC Name ethanolate;iron(3+)
SMILES CC[O-].CC[O-].CC[O-].[Fe+3]

Specifications

Melting Point 120°C
Color Brown to Gray
Boiling Point 155°C (0.1 mmHg)
Odor Odorless
Linear Formula Fe(OCH2CH3)3
Quantity 2 g
UN Number UN3181
Sensitivity Moisture sensitive
Solubility Information Soluble in anhydrous hexane,alcohols;Miscible with hexane,benzene and ethanol.
Formula Weight 191.03
Percent Purity 99.6%
Physical Form Chunks and/or Powder
Assay (metals basis)
Chemical Name or Material Iron(III) ethoxide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.