Learn More
Iron(III) ethoxide, 99.6% (metals basis)
Description
Precursor for Sol-Gel processes.Iron(III) ethoxide is used as a precursor for inorganic compound iron oxide. It is used in the preparation of iron-platinum nanoparticles by pyrolysis with platinum(II) acetylacetonate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Packaged under argon
Specifications
Specifications
| Melting Point | 120°C |
| Color | Brown to Gray |
| Boiling Point | 155°C (0.1 mmHg) |
| Odor | Odorless |
| Linear Formula | Fe(OCH2CH3)3 |
| Quantity | 10 g |
| UN Number | UN3181 |
| Sensitivity | Moisture sensitive |
| Solubility Information | Soluble in anhydrous hexane,alcohols;Miscible with hexane,benzene and ethanol. |
| Formula Weight | 191.03 |
| Show More |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.