missing translation for 'onlineSavingsMsg'
Learn More

Iron(III) citrate hydrate, Fe(III) 16.5-20%; Fe(II) max 5%

Product Code. 11474003
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
This item is not returnable. View return policy

Product Code. 11474003

Brand: Thermo Scientific Alfa Aesar A10276.0B

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 334024-15-6 | C6H5FeO7 | 244.94 g/mol

Provides a source of iron used in cell culture applications. Provides iron in a less toxic form than free iron salts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Provides a source of iron used in cell culture applications. Provides iron in a less toxic form than free iron salts.

Solubility
Soluble in water.

Notes
Light sensitive. Store away from strong oxidizing agents. keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 334024-15-6
Molecular Formula C6H5FeO7
Molecular Weight (g/mol) 244.94
MDL Number MFCD00149615
InChI Key NPFOYSMITVOQOS-UHFFFAOYSA-K
Synonym ferric citrate monohydrate, iron iii citrate hydrate, unii-334s8t6lke, ferric citrate hydrate, ferric citrate usan, iron iii citrate tribasic monohydrate, citric acid iron iii salt, iron cento citrate hydrate, ksc534c4h, iron 3+ citrate hydrate
PubChem CID 22178220
IUPAC Name 2-hydroxypropane-1,2,3-tricarboxylate;iron(3+);hydrate
SMILES [Fe+3].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O

Specifications

Quantity 1000 g
Solubility Information Soluble in water.
Formula Weight 244.94 (Anhydrous)
Concentration or Composition (by Analyte or Components) Fe(III) 16.5 to 20%; Fe(II) max 5%
Sensitivity Light sensitive
Chemical Name or Material Iron (III) citrate hydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.