missing translation for 'onlineSavingsMsg'
Learn More

Iron(III) chloride solution, Purum, 45% FeCl3 basis, Honeywell Fluka™

Product Code. 15651120 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
2.5 L
4 x 2.5 L
Unit Size:
2.5 L
Pack of 4
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15651120 4 x 2.5 L Pack of 4
15641120 2.5 L 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15651120 Supplier Honeywell-Fluka Supplier No. 123224X2.5L
Request Bulk or Custom Formats

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Purum, 45% FeCl3 basis

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7705-08-0 , 7732-18-5
Molecular Formula Cl3Fe
Molecular Weight (g/mol) 162.20
MDL Number MFCD00011005
InChI Key RBTARNINKXHZNM-UHFFFAOYSA-K
Synonym Ferric chloride solution
IUPAC Name iron(3+) trichloride
SMILES [Cl-].[Cl-].[Cl-].[Fe+3]

Specifications

CAS Min % ≥40.0000%
CAS Max % <60.0000%
Quantity 4 x 2.5 L
Linear Formula HOCH2CH2OH
UN Number UN2582
Formula Weight 162.2
Percent Purity 45%
Grade purum
Packaging Glass Bottle
Density 1.48 g/cm3
Assay FeCl3
Chemical Name or Material Iron(III) chloride solution, Purum, 45% FeCl3 basis
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.