missing translation for 'onlineSavingsMsg'
Learn More

Iron(III) chloride hexahydrate, ACS, 97.0-102.0%

Product Code. 11420290
Click to view available options
Quantity:
25 g
250 g
1 kg
5 kg
Unit Size:
1kg
250g
25g
5kg
This item is not returnable. View return policy

Product Code. 11420290

Brand: Thermo Scientific Alfa Aesar 012497.A7

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used to prepare organically templated iron phosphates.Also, used for sewerage treatment of industrial wastes, chloride hydrometallurgy. Useful in the detection of phenols and phenolic derivatives; gamma-hydroxybutyric acids and also in the Trinder spot test for detecting salicylic acids. | CAS: 10025-77-1 | Cl3FeH12O6 | 270.29 g/mol

Iron(III) chloride hexahydrate is used to prepare organically templated iron phosphates. It is used for sewerage treatment of industrial wastes, chloride hydrometallurgy (Silgarin process for the production of silicon), drinking water production, production of precursors for polyvinyl chloride (PVC) and copper-based metals in printed circuit boards. It plays an important role in photoengraving and photography as an etching agent. It is useful as a mordant in dyeing and printing textiles. It is useful in the detection of phenols and phenolic derivatives; gamma-hydroxybutyric acids and also in the Trinder spot test for detecting salicylic acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications:
  • Used to prepare organically templated Iron Phosphates
  • Used for sewerage treatment of industrial wastes, chloride hydrometallurgy (Silgarin) process for production of silicon), drinking water production, production of precursors for polyvinyl chloride (PVC) and copper-based metals in printed circuit boards
  • Plays important role in photoengraving and photography as an etching agent
  • Usedl as mordant in dyeing and printing textiles.
  • Also usedl in detection of phenols and phenol derivatives; gamma-hydroxybutyric acids, in Trinder spot test for detecting salicylic acids
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10025-77-1
Molecular Formula Cl3FeH12O6
Molecular Weight (g/mol) 270.29
MDL Number MFCD00149712
InChI Key NQXWGWZJXJUMQB-UHFFFAOYSA-K
Synonym Ferric chloride hexahydrate
IUPAC Name iron(3+) hexahydrate trichloride
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Fe+3]

Specifications

Boiling Point 280°C to 285°C
Melting Point 37°C
Color Brown-Yellow
Physical Form Lump
Quantity 5 kg
Assay Percent Range 97.0 to 102.0%
UN Number UN3260
Solubility Information Freely soluble in water, alcohol, ether, acetone.Soluble in water, alcohol, glycerol, methanol, ether and acetone. Insoluble in ethyl acetate.
Formula Weight 270.30 (162.21 Anhydrous)
Grade ACS
Odor Odorless
Sensitivity Hygroscopic
Appearance Yellow-Brown lump
Density 1.820 g/cm3
Chemical Name or Material Iron(III) Chloride Hexahydrate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Iron(III) chloride hexahydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Iron(III) chloride h

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.