Learn More
Iron(III) chloride hexahydrate, 97+%, ACS reagent, Thermo Scientific Chemicals
£22.50 - £596.00
Chemical Identifiers
CAS | 10025-77-1 |
---|---|
Molecular Formula | Cl3FeH12O6 |
Molecular Weight (g/mol) | 270.29 |
MDL Number | MFCD00149712 |
InChI Key | NQXWGWZJXJUMQB-UHFFFAOYSA-K |
Synonym | Ferric chloride hexahydrate |
IUPAC Name | iron(3+) hexahydrate trichloride |
SMILES | O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Fe+3] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10015730
|
Thermo Scientific Acros
423700050 |
5 g | Glass bottle |
£22.50
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10586982
|
Thermo Scientific Acros
423705000 |
500 g | Plastic Bottle |
£57.10
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10733164
|
Thermo Scientific Acros
423700025 |
2.5 kg | Plastic Bottle |
£186.00
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15119293
|
Thermo Scientific Acros
423700100 |
10 kg | Plastic drum |
£596.00
10kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acidChemical Identifiers
10025-77-1 | |
270.29 | |
NQXWGWZJXJUMQB-UHFFFAOYSA-K | |
iron(3+) hexahydrate trichloride |
Cl3FeH12O6 | |
MFCD00149712 | |
Ferric chloride hexahydrate | |
O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Fe+3] |
Specifications
280.0°C to 285.0°C | |
10025-77-1 | |
Crystalline Powder | |
Cl3FeH12O6 | |
MFCD00149712 | |
15,4046 | |
Solubility in water: 920g/L (20°C). Other solubilities: readily soluble in alcohol, acetone and ether | |
O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Fe+3] | |
270.29 | |
≥97% | |
Iron(III) chloride hexahydrate |
37.0°C | |
White | |
97.0 to 102% (Iodometry) | |
FeCl3·6H2O | |
01,390; 02,199; 03,145; 04,236; 05,307; 06,259; 07,153; 08,228; 09,222; 15,158 | |
Ferric chloride hexahydrate | |
NQXWGWZJXJUMQB-UHFFFAOYSA-K | |
iron(3+) hexahydrate trichloride | |
270.29 | |
ACS Reagent |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye damage.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
IF IN EYES: Rinse cautiously with wa
GHS Signal Word: Danger
missing translation for 'rtecsNumber' : NO5425000
- Iron(III) chloride hexahydrate
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 1
- Skin sensitiser Category 1
Hazard Statement
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H317-May cause an allergic skin reaction.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P313-Get medical advice/attention.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only