missing translation for 'onlineSavingsMsg'
Learn More

Iron(III) bromide, anhydrous, 98+%

Product Code. 11377408
Change view
Click to view available options
Quantity:
5 g
100 g
Unit Size:
100g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11377408 100 g 100g
11387408 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11377408 Supplier Thermo Scientific Alfa Aesar Supplier No. 031125.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10031-26-2

Iron(III) bromide is used as a Lewis acid catalyst in the halogenation of aromatic compounds. It is also used in the preparation of 2,5-dibromo-4-aryl-1,3-pentadiene by reacting with homoallenic bromohydrin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10031-26-2
MDL Number MFCD00016082

Specifications

Melting Point (decomposition)
Color Red
Physical Form Crystalline
Quantity 100 g
Merck Index 14,4018
Solubility Information Soluble in water,ether and alcohol.
Formula Weight 295.57
Percent Purity ≥98%
Odor Odorless
Sensitivity Hygroscopic; Light sensitive
Chemical Name or Material Iron(III) bromide, Anhydrous
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.