Learn More
Iron(III) bromide, 90+%, Thermo Scientific Chemicals
£124.00 - £412.00
Chemical Identifiers
CAS | 10031-26-2 |
---|---|
Molecular Formula | Br3Fe |
Molecular Weight (g/mol) | 295.56 |
MDL Number | MFCD00016082 |
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acidChemical Identifiers
10031-26-2 | |
Br3Fe | |
295.56 | |
MFCD00016082 |
Specifications
200°C | |
Beige-Yellow to White | |
90% min. (Iodometry) | |
FeBr3 | |
15, 4045 | |
295.56 | |
≥90% |
10031-26-2 | |
Powder | |
Br3Fe | |
MFCD00016082 | |
Solubility in water: soluble | |
295.56 | |
Iron(III) bromide |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 233-089-1
- Iron(III) bromide
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only