missing translation for 'onlineSavingsMsg'
Learn More

Iron(III) acrylate, 96%, Thermo Scientific™

Product Code. 11375007
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11375007 100 g 100g
11365007 5 g 5g
11355007 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11375007 Supplier Thermo Scientific Alfa Aesar Supplier No. 042001.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 55488-18-1
Molecular Formula C9H9FeO6
Molecular Weight (g/mol) 269.01
MDL Number MFCD00080380
InChI Key WNWBIDPJHFYYLM-UHFFFAOYSA-K
Synonym iron iii acrylate, feracryl, pk 2 acrylic polymer, poly acrylic acid iron salt, iron acrylate, 2-propenoic acid, homopolymer, iron salt, iron triacrylate, 79-10-7 parent, iron 3+ acrylate, 2-propenoic acid, iron 3+ salt
PubChem CID 171450
IUPAC Name iron(3+);prop-2-enoate
SMILES C=CC(=O)[O-].C=CC(=O)[O-].C=CC(=O)[O-].[Fe+3]

Specifications

Melting Point 190°C to 200°C (decomposition)
Quantity 100 g
Sensitivity Air and moisture sensitive
Formula Weight 269.01
Percent Purity 96%
Physical Form Powder
Chemical Name or Material Iron(III) acrylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.