missing translation for 'onlineSavingsMsg'
Learn More

Iron(III) 2-Ethylhexanoate (Nominally 50% in mineral spirits, Fe 6%), TRC

Product Code. 30389349
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30389349 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30389349 Supplier Toronto Research Chemicals Supplier No. TRCI7752001G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Iron(III) 2-Ethylhexanoate
CAS 7321-53-1
Molecular Formula C24H45FeO6
Quantity 1 g
Synonym 2-ethyl-Hexanoic Acid Iron(3+) Salt, Ferric 2-Ethylhexanoate, Iron Ethylhexanoate, Iron Tris(2-ethylhexanoate)
InChI Formula InChI=1S/3C8H16O2.Fe/c3*1-3-5-6-7(4-2)8(9)10;/h3*7H,3-6H2,1-2H3,(H,9,10);/q;;;+3/p-3
SMILES CCCCC(CC)C(=O)O[Fe](OC(=O)C(CC)CCCC)OC(=O)C(CC)CCCC
IUPAC Name tris(2-ethylhexanoyloxy)iron
Molecular Weight (g/mol) 485.47
Formula Weight 485.2566
Concentration or Composition (by Analyte or Components) 50 %
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.