missing translation for 'onlineSavingsMsg'
Learn More

Iron(III) 2-ethylhexanoate, nominally 50% in mineral spirits, Fe 6%, Thermo Scientific™

Product Code. 11463750
Click to view available options
Quantity:
50 g
250 g
1 kg
Unit Size:
1kg
250g
50g
This item is not returnable. View return policy

Product Code. 11463750

Brand: Thermo Scientific Alfa Aesar 089307.30

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7321-53-1
Molecular Formula C24H45FeO6
Molecular Weight (g/mol) 485.463
MDL Number MFCD00014006
InChI Key ULUYRVWYCIOFRV-UHFFFAOYSA-K
Synonym ferric 2-ethylhexanoate, iron 2-ethylhexanoate, iron tris 2-ethylhexanoate, iron ethylhexanoate, iron iii 2-ethylhexanoate, hexanoic acid, 2-ethyl-, iron 3+ salt 3:1, iron iii 2-ethylhexanoate nominally in mineral spirits, fe, iron tallate 2-ethylhexanoate, iron 3+ tris 2-ethylhexanoate, hexanoic acid, 2-ethyl-, iron 3+ salt
PubChem CID 110963
IUPAC Name 2-ethylhexanoate;iron(3+)
SMILES CCCCC(CC)C(=O)[O-].CCCCC(CC)C(=O)[O-].CCCCC(CC)C(=O)[O-].[Fe+3]

Specifications

Color Brown
Density 0.91 g/mL
Quantity 250 g
UN Number UN1268
Solubility Information Miscible with chloroform. Immiscible with water.
Formula Weight 485.47
Concentration or Composition (by Analyte or Components) Fe 6%
Physical Form Liquid
Chemical Name or Material Iron (III) 2-ethylhexanoate, Nominally 50% in mineral spirits
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.