missing translation for 'onlineSavingsMsg'
Learn More

Iron(III) 2-ethylhexanoate, nominally 50% in mineral spirits, Fe 6%, Thermo Scientific™

Product Code. 11453750
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
Unit Size:
1kg
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11453750 50 g 50g
11463750 250 g 250g
11349409 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11453750 Supplier Thermo Scientific Alfa Aesar Supplier No. 089307.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7321-53-1
Molecular Formula C24H45FeO6
Molecular Weight (g/mol) 485.463
MDL Number MFCD00014006
InChI Key ULUYRVWYCIOFRV-UHFFFAOYSA-K
Synonym ferric 2-ethylhexanoate, iron 2-ethylhexanoate, iron tris 2-ethylhexanoate, iron ethylhexanoate, iron iii 2-ethylhexanoate, hexanoic acid, 2-ethyl-, iron 3+ salt 3:1, iron iii 2-ethylhexanoate nominally in mineral spirits, fe, iron tallate 2-ethylhexanoate, iron 3+ tris 2-ethylhexanoate, hexanoic acid, 2-ethyl-, iron 3+ salt
PubChem CID 110963
IUPAC Name 2-ethylhexanoate;iron(3+)
SMILES CCCCC(CC)C(=O)[O-].CCCCC(CC)C(=O)[O-].CCCCC(CC)C(=O)[O-].[Fe+3]

Specifications

Color Brown
Density 0.91 g/mL
Quantity 50 g
UN Number UN1268
Solubility Information Miscible with chloroform. Immiscible with water.
Formula Weight 485.47
Concentration or Composition (by Analyte or Components) Fe 6%
Physical Form Liquid
Chemical Name or Material Iron (III) 2-ethylhexanoate, Nominally 50% in mineral spirits
compliance-icons
Product Identifier
  • Iron(III) 2-ethylhexanoate, nominally 50% in mineral spirits, Fe 6%
Signal Word
  • Danger
Hazard Category
  • Germ cell mutagenicity Category 1B
  • Carcinogenicity Category 1B
  • Specific target organ toxicity Category 1
  • Aspiration toxicity Category 1
Hazard Statement
  • H340-May cause genetic defects.
  • H350-May cause cancer.
  • H372-Causes damage to organs.
  • H304-May be fatal if swallowed and enters airways.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P405-Store locked up.
  • P501-Dispose of contents/container to .
Supplemental information
  • MIXTURE LIST-Contain: Mineral spirits
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.