missing translation for 'onlineSavingsMsg'
Learn More

Iron(III) 2-ethylhexanoate diisopropoxide, 10% w/v in isopropanol, Thermo Scientific™

Product Code. 11378548
Change view
Click to view available options
Quantity:
25 mL
100 mL
Unit Size:
100mL
25mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11378548 100 mL 100mL
11388548 25 mL 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11378548 Supplier Thermo Scientific Alfa Aesar Supplier No. 040583.AE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 331720-17-3
Molecular Formula C14H32FeO4
Molecular Weight (g/mol) 320.251
MDL Number MFCD00798540
InChI Key GCDBQBNMPOKFEM-UHFFFAOYSA-N
Synonym iron iii 2-ethylhexano-isopropoxide,in isopropanol, iron 3+ 2-ethylhexanoate 2-propanolate 1:1:2, iron iii ethylhexano-isopropoxide w/v in isopropanol
PubChem CID 121233720
IUPAC Name 2-ethylhexanoic acid;iron;propan-2-ol
SMILES CCCCC(CC)C(=O)O.CC(C)O.CC(C)O.[Fe]

Specifications

Quantity 100 mL
UN Number UN1219
Sensitivity Moisture sensitive
Formula Weight 317.23
Concentration or Composition (by Analyte or Components) 10% w/v in isopropanol
Physical Form Liquid
Chemical Name or Material Iron(III) 2-ethylhexanoate diisopropoxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.