Learn More
Iron(II) sulfide, 99.9% (metals basis), Thermo Scientific Chemicals
£28.40 - £164.00
Chemical Identifiers
CAS | 1317-37-9 |
---|---|
Molecular Formula | FeS |
Molecular Weight (g/mol) | 87.91 |
MDL Number | MFCD00011013 |
InChI Key | GNVXPFBEZCSHQZ-UHFFFAOYSA-N |
Synonym | ferrous sulfide, iron ii sulfide, iron sulfide, troilite, iron sulfuret, iron sulphide, iron monosulfide, iron protosulfide, ferrous monosulfide |
PubChem CID | 14828 |
SMILES | [S--].[Fe++] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11331559
|
Thermo Scientific Alfa Aesar
014024.03 |
1 g |
£28.40
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11341559
|
Thermo Scientific Alfa Aesar
014024.09 |
10 g |
£49.20
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11351559
|
Thermo Scientific Alfa Aesar
014024.18 |
50 g |
£164.00
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Iron(II) sulfide is used to generate hydrogen sulfide instantaneously by reacting with hydrochloric acid. In laboratory, Kipp’s apparatus is utilized for this purpose. It is used as a re-sulphurizing and alloying agent, as a reducing agent to remove heavy metal impurities from phosphoric acid. It is used to control hydrogen embrittlement in alloy and stainless steel industries.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1317-37-9 | |
87.91 | |
GNVXPFBEZCSHQZ-UHFFFAOYSA-N | |
14828 |
FeS | |
MFCD00011013 | |
ferrous sulfide, iron ii sulfide, iron sulfide, troilite, iron sulfuret, iron sulphide, iron monosulfide, iron protosulfide, ferrous monosulfide | |
[S--].[Fe++] |
Specifications
1195°C | |
Brown to Black | |
99.9% (Metals basis) | |
MFCD00011013 | |
Insoluble in water and nitric acid. | |
[S--].[Fe++] | |
87.91 | |
87.91 | |
Air and moisture sensitive | |
Iron(II) sulfide |
1317-37-9 | |
Powder | |
FeS | |
ferrous sulfide, iron ii sulfide, iron sulfide, troilite, iron sulfuret, iron sulphide, iron monosulfide, iron protosulfide, ferrous monosulfide | |
GNVXPFBEZCSHQZ-UHFFFAOYSA-N | |
sulfanylideneiron | |
14828 | |
Odorless | |
4.74 |
Safety and Handling
This material and/or its container must be disposed of as hazardous waste.
Contact with acids liberates toxic gas.
missing translation for 'einecsNumber' : 215-268-6
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only