missing translation for 'onlineSavingsMsg'
Learn More

Iron(II) selenide, 99.9% (metals basis)

Product Code. 11347398
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11347398 10 g 10g
11337398 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11347398 Supplier Thermo Scientific Alfa Aesar Supplier No. 031112.09
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 1310-32-3 | FeSe | 134.82 g/mol

Iron(II) selenide is used in electrical semiconductors. It is transformed into superconductor by a purely chemical method.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1310-32-3
Molecular Formula FeSe
Molecular Weight (g/mol) 134.82
MDL Number MFCD00016085
InChI Key WALCGGIJOOWJIN-UHFFFAOYSA-N
Synonym iron selenide, iron ii selenide
PubChem CID 14795
IUPAC Name selanylideneiron
SMILES [Fe]=[Se]

Specifications

Color Black
Physical Form Powder
Quantity 10 g
Assay Percent Range 99.9% (Metals basis)
Solubility Information Insoluble in water.
Formula Weight 134.81
Odor Odorless
Sensitivity Moisture sensitive
Density 6.9g/cm3
Chemical Name or Material Iron(II) selenide
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Iron(II) selenide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Specific target organ toxicity after repeated exposure Category 2
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H301-Toxic if swallowed.
  • H331-Toxic if inhaled.
  • H373-May cause damage to organs.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to accordance with local/regional/national/international regulations..

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.