missing translation for 'onlineSavingsMsg'
Learn More

Iron(II) oxide, 99.5% (metals basis)

Product Code. 11357288
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11357288 25 g 25g
11367288 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11357288 Supplier Thermo Scientific Alfa Aesar Supplier No. 030513.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

In manufacture of green, heat-absorbing glass | CAS: 1345-25-1 | FeO | 71.844 g/mol

In manufacture of green, heat-absorbing glass; in steel manufacture; and as a catalystIron(II) oxide is used in manufacturing of green, heat-absorbing glass; in steel manufacture; and as a catalyst. It is used as a dye or pigment in pottery, glazes and glasses. It finds application as heat-absorbing glass used in buildings and automobiles. It is used as a precursor in the production of steel. It is also used in cosmetics and some tattoo inks. It can also be used for filtering phosphates from home aquaria and acts as a catalyst in a number of industrial and chemical operations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1345-25-1
Molecular Formula FeO
Molecular Weight (g/mol) 71.844
MDL Number MFCD00016095
InChI Key UQSXHKLRYXJYBZ-UHFFFAOYSA-N
Synonym iron ii oxide, ferrous oxide, iron monooxide, iron monoxide, iron oxide, wustite, natural wuestite, eisenoxydul, hsdb 464, eisen ii-oxid
PubChem CID 14945
ChEBI CHEBI:50820
IUPAC Name oxoiron
SMILES O=[Fe]

Specifications

Melting Point 1369°C
Quantity 25 g
Assay Percent Range 99.5% (metals basis)
Solubility Information Insoluble in water and alkalies. Soluble in HCl,H2SO4. Slightly soluble in HNO3Insoluble in water,alcohol and alkali. Soluble in hydrochloric acid and sulfuric acid. Slightly soluble in nitric acid.
Formula Weight 71.85
Sensitivity Air Sensitive
Density 5.7
Chemical Name or Material Iron(II) oxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.