missing translation for 'onlineSavingsMsg'
Learn More

Iron(II) oxalate hydrate, 96%

Product Code. 11373658
Change view
Click to view available options
Quantity:
500 g
2 kg
Unit Size:
2kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11373658 500 g 500g
11383658 2 kg 2kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11373658 Supplier Thermo Scientific Alfa Aesar Supplier No. 039261.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 516-03-0 | C2FeO4 | 143.86 g/mol

Used as a photographic developer, glass tint, decorative glass colorant, and pigment in plastics, paints, and lacquers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 516-03-0
Molecular Formula C2FeO4
Molecular Weight (g/mol) 143.86
MDL Number MFCD00012475
InChI Key OWZIYWAUNZMLRT-UHFFFAOYSA-L
Synonym ferrous oxalate, iron ii oxalate, unii-dzp4yv3icv, dzp4yv3icv, iron protoxalate, iron, ethanedioato 2--.kappa.o1,.kappa.o2, iron 2+ oxalate, iron oxalate fec2o4, hsdb 463, iron 2+ ; oxalate
PubChem CID 10589
IUPAC Name iron(2+);oxalate
SMILES [Fe++].[O-]C(=O)C([O-])=O

Specifications

Melting Point 190°C (decomposition)
Density 2.28 g/mL
Quantity 500 g
UN Number UN3288
Sensitivity Hygroscopic
Solubility Information Soluble in acids
Formula Weight 143.87 (Anhydrous)
Percent Purity 96%
Physical Form Powder
Chemical Name or Material Iron(II) oxalate hydrate
compliance-icons
Product Identifier
  • Iron(II) oxalate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P363-Wash contaminated clothing before reuse.
  • P321-Specific treatment (see on this label).
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P330-Rinse mouth.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P501-Dispose of contents/container to .

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.