Learn More
Iron(II) oxalate dihydrate, Puratronic™, 99.999% (metals basis)
Description
It is used as photographic developer for silver bromide-gelatin plates. It imparts greenish-brown tint to optical glass (sunglasses, windshields, railroad car windows), for decorative glassware, pigment for plastics, paints and lacquers. It acts as reagent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 190°C (decomposition) |
| Physical Form | Powder |
| Quantity | 2.5 g |
| Solubility Information | Soluble in water. Insoluble in acetic acid. |
| IUPAC Name | iron(2+);oxalate;dihydrate |
| Formula Weight | 179.90 (143.87 Anhydrous) |
| Percent Purity | 99.999% |
| Grade | Puratronic |
| Sensitivity | Hygroscopic |
| Density | 2.28 g/mL |
| Show More |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.