missing translation for 'onlineSavingsMsg'
Learn More

Iron(II) methoxide, 98%

Product Code. 11358057
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11358057 10 g 10g
11368057 2 g 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11358057 Supplier Thermo Scientific Alfa Aesar Supplier No. 022552.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7245-21-8 | C2H6FeO2 | 117.913 g/mol

Iron(II) methoxide, is used in Engineering & Environmental, Catalysis & Synthesis, and In organic syntheses, chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7245-21-8
Molecular Formula C2H6FeO2
Molecular Weight (g/mol) 117.913
MDL Number MFCD00061474
InChI Key VAPRHKOWFRYFTF-UHFFFAOYSA-N
Synonym iron ii methoxide, iron 2+ dimethoxide, methanol, iron 2+ salt, acmc-1bc40
PubChem CID 14598007
IUPAC Name iron(2+);methanolate
SMILES C[O-].C[O-].[Fe+2]

Specifications

Color Green
Odor Alcohol-like
Linear Formula Fe(OCH3)2
Quantity 10 g
UN Number UN3181
Sensitivity Air and moisture sensitive
Solubility Information Reacts with water.
Formula Weight 117.92
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Iron(II) methoxide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.